missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phthalimide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Phthalimide potassium salt |
| CAS | 1074-82-4 |
| Molecular Formula | C8 H4 N O2 . K |
| Quantity | 25 g |
| Synonym | 1H-Isoindole-1,3(2H)-dione','potassium salt (1:1) (ACI), 1H-Isoindole-1,3(2H)-dione','potassium salt (9CI), Phthalimide','potassium deriv. (6CI), Phthalimide','potassium salt (8CI), Potassium','phthalimido- (7CI), 1H-Isoindole-1,3(2H)-dione','potassium salt','ion(1-) (1:1), Isoindole-1,3-dione potassium salt, N-Potassiophthalimide, Phthalimide potassium, Phthalimidopotassium, Potassium 1,3-dioxoisoindolin-2-ide, Potassium 2,3-dihydro-1H-isoindole-1,3-dione, Potassium isoindoline-1,3-dione, Potassium phthalimidate, Potassium phthalimide |
| InChI Formula | InChI=1S/C8H5NO2.K/c10-7-5-3-1-2-4-6(5)8(11)9-7;/h1-4H,(H,9,10,11);/q;+1/p-1 |
| SMILES | [K+].O=C1[N-]C(=O)c2ccccc12 |
| IUPAC Name | potassium;isoindol-2-ide-1,3-dione |
| Molecular Weight (g/mol) | 185.22 |
| Formula Weight | 184.9879 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction