missing translation for 'onlineSavingsMsg'
Learn More

Potassium pyrazole-5-trifluoroborate, 95%, Thermo Scientific™

Product Code. 15454817
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
This item is not returnable. View return policy

Product Code. 15454817

missing translation for 'mfr': Thermo Scientific Alfa Aesar H32128.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013640-87-3
Molecular Formula C3H3BF3KN2
Molecular Weight (g/mol) 173.975
MDL Number MFCD09992890
InChI Key QNHYBMZHGRWURK-UHFFFAOYSA-N
Synonym potassium 1h-pyrazole-5-trifluoroborate, potassium pyrazole-5-trifluoroborate, potassium 1h-pyrazole-3-trifluoroborate, potassium trifluoro 2h-pyrazol-3-yl boranuide, potassium trifluoro 1h-pyrazol-3-yl borate, potassium trifluoro 1h-pyrazol-5-yl borate 1-, potassium trifluoro 1h-pyrazol-3-yl boranuide
PubChem CID 25212877
IUPAC Name potassium;trifluoro(1H-pyrazol-5-yl)boranuide
SMILES [B-](C1=CC=NN1)(F)(F)F.[K+]

Specifications

Quantity 250 mg
Formula Weight 173.98
Percent Purity 95%
Chemical Name or Material Potassium pyrazole-5-trifluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.