missing translation for 'onlineSavingsMsg'
Learn More

Potassium pyrophosphate, 95%

Product Code. 11386768
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Product Code. Quantity unitSize
11386768 1 kg 1kg
11376768 5 kg 5kg
2 options
This item is not returnable. View return policy

Product Code. 11386768

Brand: Thermo Scientific Alfa Aesar 013436.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7320-34-5 | K4O7P2 | 330.33 g/mol

Potassium pyrophosphate is mainly used as an emulsifier, quality improver and metal ion chelating agent. It can be found in herbicidal compositions as a spray adjuvant. It is also used as an anionic dispersant in water-based (latex) paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7320-34-5
Molecular Formula K4O7P2
Molecular Weight (g/mol) 330.33
MDL Number MFCD00011393
InChI Key RYCLIXPGLDDLTM-UHFFFAOYSA-J
Synonym potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate
PubChem CID 23740
IUPAC Name tetrapotassium;phosphonato phosphate
SMILES [K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Boiling Point (decomposition)
Melting Point 1090°C
Color White
Physical Form Powder
Quantity 1 kg
Merck Index 14,7663
Solubility Information Soluble in water
Formula Weight 330.34
Percent Purity 95%
Sensitivity Hygroscopic
Density 2.33 g/mL
Chemical Name or Material Potassium pyrophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium pyrophosphate, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tetrapotassium pyrophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.