missing translation for 'onlineSavingsMsg'
Learn More

Potassium Pyrophosphate, MP Biomedicals

Product Code. 11408215 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408215 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408215 Supplier MP Biomedicals Supplier No. 0521755980
 Restricted Item
Estimated Shipment: 18-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical reagent for research and manufacturing use.

Specifications

CAS 7320-34-5
Physical Form Powder
Quantity 100 g
Molecular Formula K4O7P2
MDL Number MFCD00011393
Synonym potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate
InChI Key RYCLIXPGLDDLTM-UHFFFAOYSA-J
SMILES [K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O
IUPAC Name tetrapotassium (phosphonatooxy)phosphonate
Molecular Weight (g/mol) 330.33
PubChem CID 23740
Formula Weight 384.4
Percent Purity 98.73%
Chemical Name or Material Potassium Pyrophosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.