missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium selenocyanate, 99%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 152.0°C to 158.0°C |
| Color | Orange to Red |
| Physical Form | Crystalline Solid, Powder, Crystals and/or Chunks |
| Quantity | 10 g |
| Assay Percent Range | 98.5 % min. (Argentometry) |
| Linear Formula | KSeCN |
| Fieser | 06,487; 08,417; 13,186; 08,417; 15,274 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 144.08 |
| Percent Purity | 99% |
| Show More |
Seguridad y manipulación
Nombre del producto
- Potassium selenocyanate
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido