missing translation for 'onlineSavingsMsg'
Learn More

Potassium sodium tartrate tetrahydrate, 99+%, for analysis

Product Code. 10638343 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Drum
Plastic bottle
Unit Size:
1kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10638343 5 kg Plastic Drum 5kg
10462011 500 g Plastic bottle 500g
10721861 1 kg Plastic bottle 1kg
15109233 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10638343 Supplier Thermo Scientific Acros Supplier No. 202860051
  • ONLINE EXCLUSIVE PRICE
    316.00 €
    306.00 € / 5kg
    Save 10.00 €
    3% Off
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6381-59-5 | C4H4KNaO6·4H2O | 282.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-59-5
MDL Number MFCD00150989
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Decomposition Information >100°C
Melting Point 70.0°C to 80.0°C
Color White
pH 5.5 to 8.5 (5% soln. at 20°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic Drum
Molecular Formula C4H4KNaO6·4H2O
Linear Formula KO2CCH(OH)CH(OH)CO2Na·4H2O
Quantity 5 kg
Beilstein 03, 525
Merck Index 15, 7790
Specific Rotation Condition + 22.10 (20.00°C c=10,water)
Specific Rotation + 22.10
Solubility Information Solubility in water: 630g/L (20°C). Other solubilities: practically insoluble in alcohol
Molecular Weight (g/mol) 282.22
Formula Weight 282.22
Percent Purity 99+%
Grade Analysis
Physical Form Crystals
Chemical Name or Material Potassium sodium tartrate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.