missing translation for 'onlineSavingsMsg'
Learn More

Potassium sodium tartrate tetrahydrate, ACS reagent

Product Code. 12643600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12643600 2.5 kg 2.5kg
12675067 100 g 100g
12685067 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12643600 Supplier Thermo Scientific Acros Supplier No. 447220025
  • ONLINE EXCLUSIVE PRICE
    283.00 €
    265.00 € / 2.5kg
    Save 18.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6381-59-5 | C4H4KNaO6·4H2O | 282.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H4KNaO6·4H2O
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Melting Point 70.0°C to 80.0°C
Quantity 2.5 kg
Packaging Plastic bottle
Linear Formula KO2CCH(OH)CH(OH)CO2Na·4H2O
Merck Index 15,779
Solubility Information Solubility in water: 630g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 282.22
Grade ACS Reagent
Chemical Name or Material Potassium sodium tartrate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.