missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sodium Tartrate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10539010 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10539010 500 g Plastic powder jar 500g
12636757 1 kg Plastic powder jar 1kg
12646757 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10539010 Supplier Fisher Chemical Supplier No. P/6800/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.218
MDL Number 150989
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Melting Point 70°C
Color White
pH 5.5 to 8.5
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 282.22g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.