missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sodium Tartrate Tetrahydrate, Extra Pure, SLR, Powder, Fisher Chemical

Product Code. 10519200 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10519200 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10519200 Supplier Fisher Chemical Supplier No. P/6840/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 70°C
CAS 6381-59-5
Color White
pH 5.5 to 8.5
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C4H12KNaO10
MDL Number 150989
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
Molecular Weight (g/mol) 282.218
PubChem CID 2724148
Formula Weight 282.22g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.