missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, 99.99% (metals basis)

Product Code. 11327288
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11327288 5 g 5g
11307288 25 g 25g
11317288 100 g 100g
3 options
This item is not returnable. View return policy

Artikelnummer. 11327288

missing translation for 'mfr': Thermo Scientific Alfa Aesar 030485.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate, and glass. In analytical chemistry, it is used in the determination of nitrogen by Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is also used in the separation of protein through electrophoresis. It also serves as a laboratory reagent..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Spezifikation

Boiling Point 1689°C
Melting Point 1,069°C
Physical Form Powder
Quantity 5 g
Merck Index 14,7674
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide.
IUPAC Name dipotassium sulfate
Formula Weight 174.27
Percent Purity 99.99%
Odor Odorless
Density 2.662 g/mL
Assay (metals basis)
Chemical Name or Material Potassium sulfate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.