missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, 99+%, for analysis, anhydrous powder

Product Code. 10052720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic bottle
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10052720 500 g Plastic bottle 500g
10502364 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10052720 Supplier Thermo Scientific Acros Supplier No. 207775000
  • ONLINE EXCLUSIVE PRICE
    35.85 €
    34.80 € / 500g
    Save 1.05 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689.0°C
Melting Point 1067.0°C
Color White
pH 6 to 8 (5% soln. at 20°C)
Physical Form Very Fine Crystals or Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula K2SO4
Merck Index 15, 7793
Solubility Information Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol
IUPAC Name dipotassium;sulfate
Formula Weight 174.25
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 2.66
Chemical Name or Material Potassium sulfate, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.