missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, (Powder/ACS Reagent Grade), MP Biomedicals

Product Code. 11457852 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457852 500 g 500g
11467852 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457852 Supplier MP Biomedicals Supplier No. 0215256490
  • ONLINE EXCLUSIVE PRICE
    37.65 €
    35.70 € / 500g
    Save 1.95 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 500 g
IUPAC Name dipotassium;sulfate
Formula Weight 174.26
Grade ACS Reagent
Chemical Name or Material Potassium Sulfate, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.