missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99%, Solstice

Product Code. 15693360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
50 kg
6 x 1 kg
Unit Size:
1kg
50kg
5kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15693360 6 x 1 kg Pack of 6
15663360 1 kg 1kg
15683360 5 kg 5kg
15673360 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15693360 Supplier Honeywell-Fluka Supplier No. 312706X1KG
  • ONLINE EXCLUSIVE PRICE
    440.00 €
    241.97 € / Pack of 6
    Save 198.03 €
    45% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1,698°C
CAS Max % 100.0000%
Physical Form Crystalline Powder
Quantity 6 x 1 kg
Linear Formula K2SO4
IUPAC Name dipotassium;sulfate
Formula Weight 174.26
Percent Purity >99%
Grade ACS Reagent/Reagent ISO
Packaging Poly Bottle
Density 2.66 g/cm3
Chemical Name or Material Potassium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.