missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulphate, TRC

Product Code. 30415499
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415499 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415499 Supplier Toronto Research Chemicals Supplier No. TRCP69830550G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Sulphate
CAS 7778-80-5
Quantity 50 g
Molecular Formula K2O4S
Synonym Sulfuric Acid Dipotassium Salt, Potassium Sulfate, Arcanum Duplicatum, Dipotassium Sulfate, Potassium Sulfate, Potassium Sulfate (K2(SO4)), Potassium Sulfate (K2SO4), Potassium Sulphate, Sal Polychrestum, Sulfate of Potash, Sulfuric Acid Dipotassium Salt, Tartarus Vitriolatus
InChI Formula InChI=1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
SMILES [K+].[K+].[O-]S(=O)(=O)[O-]
IUPAC Name dipotassium;sulfate
Molecular Weight (g/mol) 174.26
Formula Weight 173.8791
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.