missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™

Product Code. 12675007 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12675007 100 mL AcroSeal™ Glass Bottle 100mL
12685007 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12675007 Supplier Thermo Scientific Acros Supplier No. 445831000
  • ONLINE EXCLUSIVE PRICE
    219.00 €
    212.00 € / 100mL
    Save 7.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium tert-butoxide
CAS 865-47-4, 96-47-9
Quantity 100 mL
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Flash Point -11°C
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
Solubility Information Solubility in water: reacts
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES CC(C)(C)[O-].[K+]
IUPAC Name potassium;2-methylpropan-2-olate
Molecular Weight (g/mol) 112.21
PubChem CID 23665647
Formula Weight 112.21
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.86
Density 0.8600g/mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402-Store in a dry place.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.