missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF, AcroSeal™

Product Code. 12685007 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12685007 800 mL AcroSeal™ Glass Bottle 800mL
12675007 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12685007 Supplier Thermo Scientific Acros Supplier No. 445838000
  • ONLINE EXCLUSIVE PRICE
    575.00 €
    534.00 € / 800mL
    Save 41.00 €
    7% Off
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Potassium tert-butoxide
CAS 865-47-4, 96-47-9
Quantity 800 mL
Flash Point -11°C
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.86
Density 0.8600g/mL
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
Solubility Information Solubility in water: reacts
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES CC(C)(C)[O-].[K+]
IUPAC Name potassium;2-methylpropan-2-olate
Molecular Weight (g/mol) 112.21
PubChem CID 23665647
Formula Weight 112.21
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Potassium tert-butoxide, 2M (25% w/w) solution in 2-MeTHF
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Self-heating substance Category 2
  • Skin corrosion/irritation Category 1 A
Gefahrenhinweis
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H252-Self-heating in large quantities; may catch fire.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.