missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-pentyloxide, 25% w/w in toluene

Product Code. 11308146
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 11308146

Brand: Thermo Scientific Alfa Aesar L15111.AU

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41233-93-6 | C5H11KO | 126.24 g/mol

Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.

Notes
Air and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and acids. Reacts violently with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41233-93-6
Molecular Formula C5H11KO
Molecular Weight (g/mol) 126.24
MDL Number MFCD00064808
InChI Key ZRLVQFQTCMUIRM-UHFFFAOYSA-N
Synonym potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
PubChem CID 23683543
IUPAC Name potassium;2-methylbutan-2-olate
SMILES [K+].CCC(C)(C)[O-]

Specifications

Density 0.87
Flash Point 4°C (39°F)
Quantity 1000 mL
UN Number UN2924
Beilstein 4149025
Sensitivity Air and moisture sensitive
Formula Weight 126.24
Concentration or Composition (by Analyte or Components) 25% w/w in toluene
Chemical Name or Material Potassium tert-pentyloxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.