missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrabromoaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 33% min

Product Code. 11357497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357497 5 g 5g
11347497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357497 Supplier Thermo Scientific Alfa Aesar Supplier No. 012664.06
 Restricted Item
Estimated Shipment: 21-12-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 13005-38-4 | KAuBr4·2H2O

It is used as a Laboratory chemical and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 13005-38-4
Molecular Formula KAuBr4·2H2O
MDL Number MFCD00049650
Synonym Gold potassium bromide

Spécification

Color Red to Brown
Physical Form Crystalline
Quantity 5 g
Odor Weak
Appearance Red-brown crystals
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water and alcohol
Formula Weight 591.72 (555.69 Anhydrous)
Chemical Name or Material Potassium tetrabromoaurate(III) dihydrate, Au 33% min.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.