missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroaurate(III) hydrate, 99%

Product Code. 11450897
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450897 1 g 1g
11460897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450897 Supplier Thermo Scientific Alfa Aesar Supplier No. B23292.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27988-75-6 | AuCl4K | 377.86 g/mol

Potassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.

Solubility
Soluble in water, ethanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27988-75-6
Molecular Formula AuCl4K
Molecular Weight (g/mol) 377.86
MDL Number MFCD00043088
InChI Key IKHCGJWURGSQRT-UHFFFAOYSA-J
Synonym potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion
PubChem CID 91886546
IUPAC Name potassium;tetrachlorogold(1-);hydrate
SMILES [K+].Cl[Au+3](Cl)(Cl)Cl

Specifications

Melting Point 357°C (decomposition)
Quantity 1 g
Linear Formula KAuCl4·xH2O
UN Number UN3260
Merck Index 14,7681
Solubility Information Soluble in water,ethanol.
Formula Weight 377.88 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Potassium tetrachloroaurate(III) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.