missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloropalladate(II), min 32.0% Pd

Product Code. 10164070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10164070 10 g Glass bottle 10g
10002560 2 g Glass bottle 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10164070 Supplier Thermo Scientific Acros Supplier No. 195120100
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10025-98-6 | Cl4K2Pd | 326.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-98-6
Molecular Formula Cl4K2Pd
Molecular Weight (g/mol) 326.41
MDL Number MFCD00011373
InChI Key LGCKLDWLSVFMGL-UHFFFAOYSA-J
Synonym potassium tetrachloropalladate ii, potassium chloropalladite, potassium tetrachloropalladate, ccris 6625, potassium chlorophalladite, potassium palladium chloride, potassium palladous chloride, dipotassium tetrachloropalladate ii, dipotassium tetrachloropalladate 2-, tetrachloropalladate 2-dipotassium
PubChem CID 61438
IUPAC Name dipotassium;tetrachloropalladium(2-)
SMILES Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]

Specifications

Melting Point 105.0°C
Color Brown to Brown-Green
Physical Form Crystalline Powder and/or Crystals
Quantity 10 g
Packaging Glass bottle
Specific Gravity 2.67
Density 2.6700g/mL
Linear Formula K2PdCl4
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 326.41
Percent Purity ≥32% (Pd)
Chemical Name or Material Potassium tetrachloropalladate(II), min 32.0% Pd
Show More Show Less
compliance-icons
Product Identifier
  • Potassium tetrachloropalladate(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.