missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate(II), 46-47% Pt

Product Code. 10144260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10144260 5 g Glass bottle 5g
10594131 1 g Glass bottle 1g
10348753 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10144260 Supplier Thermo Scientific Acros Supplier No. 195360050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-99-7
Molecular Formula Cl4H4K2Pt
Molecular Weight (g/mol) 419.11
MDL Number MFCD00011378
InChI Key JCAXKWJYKUEOFO-UHFFFAOYSA-N
Synonym potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
PubChem CID 61440
SMILES Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

Specifications

Melting Point 250.0°C
Color Red to Brown
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Linear Formula K2PtCl4
Fieser 02,182; 04,215
Merck Index 15, 7801
Solubility Information Solubility in water: 10g/L (20°C)
IUPAC Name dipotassium tetrachloroplatinumdiuide
Formula Weight 415.1
Percent Purity 46 to 47% Pt
Packaging Glass bottle
Chemical Name or Material Potassium tetrachloroplatinate(II)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.