missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tetracyanoborate, TRC

Product Code. 30426208
Change view
Click to view available options
Quantity:
2 mg
4 mg
20 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30426208 20 mg
30425164 2 mg
30418172 4 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426208 Supplier Toronto Research Chemicals Supplier No. TRCP99361820MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261356-49-4
Molecular Formula C4 B N4 . K
Molecular Weight (g/mol) 153.979
IUPAC Name potassium;tetracyanoboranuide
SMILES [K+].N#C[B-](C#N)(C#N)C#N

Specifications

Quantity 20 mg
InChI Formula InChI=1S/C4BN4.K/c6-1-5(2-7,3-8)4-9;/q-1;+1
Formula Weight 153.985
Chemical Name or Material Potassium Tetracyanoborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.