missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrafluoroborate, 99%, extra pure

Product Code. 10780561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10780561 25 g Plastic bottle 25g
10503801 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10780561 Supplier Thermo Scientific Acros Supplier No. 269220250
  • ONLINE EXCLUSIVE PRICE
    37.30 €
    35.00 € / 25g
    Save 2.30 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14075-53-7 | BF4K | 125.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14075-53-7
Molecular Formula BF4K
Molecular Weight (g/mol) 125.90
MDL Number MFCD00011395
InChI Key AKEBROIVCDHVSD-UHFFFAOYSA-N
Synonym potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
PubChem CID 518872
IUPAC Name potassium tetrafluoroboranuide
SMILES [K+].F[B-](F)(F)F

Specifications

Melting Point 530.0°C
Color White to Gray
Physical Form Micro-crystalline Powder
Quantity 25 g
Packaging Plastic bottle
Assay Percent Range 98.5% min
Density 2.5
Linear Formula KBF4
Merck Index 15,7804
Solubility Information Solubility in water: 4.4g/L (20°C). Other solubilities: slightly soluble in boiling alcohol
Formula Weight 125.91
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Potassium tetrafluoroborate, c.p.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.