missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrafluoroborate, 99%

Product Code. 11397267
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397267 2 kg 2kg
11307277 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397267 Supplier Thermo Scientific Alfa Aesar Supplier No. 011556.A3
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14075-53-7 | BF4K | 125.90 g/mol

Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as an additive in the alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14075-53-7
Molecular Formula BF4K
Molecular Weight (g/mol) 125.90
MDL Number MFCD00011395
InChI Key AKEBROIVCDHVSD-UHFFFAOYSA-N
Synonym potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
PubChem CID 518872
IUPAC Name potassium;tetrafluoroborate
SMILES [K+].F[B-](F)(F)F

Specifications

Boiling Point (decomposition)
Melting Point 529°C
Physical Form Powder
Quantity 2 kg
Linear Formula KBF4
Merck Index 14,7686
Solubility Information Soluble in water (0;45g/100g H2O); Slightly soluble in boiling alcoholSlightly soluble in water and hot ethyl alcohol; Insoluble in cold ethyl alcohol.
Formula Weight 125.91
Percent Purity ≤99%
Refractive Index 1.3245
Sensitivity Hygroscopic
Density 2.50 g/mL
Assay Typically >99.5%
Chemical Name or Material Potassium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.