missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrakis(4-chlorophenyl)borate, 98%

Product Code. 11307515
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307515 250 mg 250mg
11397505 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307515 Supplier Thermo Scientific Alfa Aesar Supplier No. L05609.MD
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14680-77-4 | C24H16BCl4K | 496.10 g/mol

Potassium tetrakis(4-chlorophenyl)borate is used as lipophilic anionic site in constructing fluorescent optode for sodium. It was also used as coating for the piezoelectric crystal in quantifying Potassium using quartz crystal microbalance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrakis(4-chlorophenyl)borate is used as lipophilic anionic site in constructing fluorescent optode for sodium. It was also used as coating for the piezoelectric crystal in quantifying Potassium using quartz crystal microbalance.

Solubility
Soluble in chloroform.

Notes
Hygroscopic. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14680-77-4
Molecular Formula C24H16BCl4K
Molecular Weight (g/mol) 496.10
MDL Number MFCD00043105
InChI Key VHPYLDPFVZBNQT-UHFFFAOYSA-N
Synonym potassium tetrakis 4-chlorophenyl borate, tetrakis 4-chlorophenyl boron potassium, k-tcpb, potassium tetrakis 4-chlorophenyl boranuide, potassium tetrakis 4-chlorophenyl borate, selectophore tm
PubChem CID 23679691
IUPAC Name potassium;tetrakis(4-chlorophenyl)boranuide
SMILES [B+3].[K+].ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 4117827
Solubility Information Soluble in chloroform.
Formula Weight 496.12
Percent Purity 98%
Chemical Name or Material Potassium tetrakis(4-chlorophenyl)borate
compliance-icons
Product Identifier
  • Potassium tetrakis(4-chlorophenyl)borate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium tetrakis(4-chlorophenyl)borate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.