missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetraphenylborate, 97%

Product Code. 15461478
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461478 5 g 5g
15471478 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461478 Supplier Thermo Scientific Alfa Aesar Supplier No. H55276.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3244-41-5 | C24H20BK | 358.332 g/mol

It is an active pharmaceutical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intemediate.

Solubility
Low solubility in water. Soluble in organic solvents.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3244-41-5
Molecular Formula C24H20BK
Molecular Weight (g/mol) 358.332
MDL Number MFCD03093877
InChI Key MAMCUZQNCMYPCY-UHFFFAOYSA-N
Synonym potassium tetraphenylborate, potassium tetraphenylboron, potassium tetraphenylboride, potassium tetrakis phenyl borate, potassium tetraphenyl boron, potassium tetraphenylborate 97, potassium tetraphenylborate 1-
PubChem CID 23665558
IUPAC Name potassium;tetraphenylboranuide
SMILES [B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[K+]

Specifications

Melting Point >300°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula KB(C6H5)4
Solubility Information Low solubility in water. Soluble in organic solvents.
Formula Weight 358.33
Percent Purity 97%
Chemical Name or Material Potassium tetraphenylborate
compliance-icons
Product Identifier
  • Potassium tetraphenylborate, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium tetraphenylborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.