missing translation for 'onlineSavingsMsg'
Learn More

Potassium thioacetate, 98%

Product Code. 10318360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10318360 25 g Glass bottle 25g
10104660 100 g Glass bottle 100g
11975731 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10318360 Supplier Thermo Scientific Acros Supplier No. 221300250
  • ONLINE EXCLUSIVE PRICE
    88.10 €
    85.50 € / 25g
    Save 2.60 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10387-40-3 | C2H3KOS | 114.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10387-40-3
Molecular Formula C2H3KOS
Molecular Weight (g/mol) 114.2
InChI Key SDJHDRMYZQFJJO-UHFFFAOYSA-N
Synonym potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
PubChem CID 24201352
IUPAC Name ethanethioic S-acid;potassium
SMILES CC(=O)S.[K]

Specifications

Melting Point 173.0°C to 176.0°C
Color White to Brown
pH ≤11 (10% aq. soln.)
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 98%
Linear Formula CH3COSK
Beilstein 02,23
Fieser 01,955
Solubility Information Solubility in water: freely soluble in water
Formula Weight 114.2
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Appearance (10% in water) clear
Chemical Name or Material Potassium thioacetate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium thioacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.