missing translation for 'onlineSavingsMsg'
Learn More

Potassium thioacetate, 98%

Product Code. 11327435
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327435 100 g 100g
11317435 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327435 Supplier Thermo Scientific Alfa Aesar Supplier No. L05405.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10387-40-3 | C2H4KOS | 115.211 g/mol

Potassium thioacetate is used for palladium mediated coupling with aryl halides and triflates leading to S-arylthioacetates and derivatives. It is also used as a reagent in the conversion of halides to thiols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thioacetate is used for palladium mediated coupling with aryl halides and triflates leading to S-arylthioacetates and derivatives. It is also used as a reagent in the conversion of halides to thiols.

Solubility
Soluble in water, methanol and some polar solvents. Insoluble in alkanes, ethers and most nonpolar solvents.

Notes
Air sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10387-40-3
Molecular Formula C2H4KOS
Molecular Weight (g/mol) 115.211
MDL Number MFCD00083065
InChI Key SDJHDRMYZQFJJO-UHFFFAOYSA-N
Synonym potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
PubChem CID 24201352
IUPAC Name ethanethioic S-acid;potassium
SMILES CC(=O)S.[K]

Specifications

Melting Point ∽240°C (decomposition)
Odor Stench
Quantity 100 g
UN Number UN3335
Beilstein 3595448
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water,methanol and some polar solvents. Insoluble in alkanes,ethers and most nonpolar solvents.
Formula Weight 114.21
Percent Purity 98%
Chemical Name or Material Potassium thioacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.