missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thioacetate, TRC

Product Code. 30420269
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420269 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420269 Supplier Toronto Research Chemicals Supplier No. TRCP698500250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Thioacetate
CAS 10387-40-3
Molecular Formula C2H4KOS
Quantity 250 g
Synonym Thioacetic Acid Potassium Salt, Ethanethioic Acid Potassium Salt, MeCOSK, Potassium Ethanethioate, Potassium Thioethanoate, Potassium Thiolacetate, S-Potassium Thioacetate
InChI Formula InChI=1S/C2H4OS.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1
SMILES [K+].CC(=O)[S-]
IUPAC Name potassium;ethanethioate
Molecular Weight (g/mol) 114.21
Formula Weight 113.9542
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.