missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 0.1N Standardized Solution

Product Code. 11393707
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393707 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11393707 Supplier Thermo Scientific Alfa Aesar Supplier No. 035602.K2
 Restricted Item
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 333-20-0 | CKNS | 97.176 g/mol

Potassium thiocyanate, 0.1N standardized solution is used in analytical chemistry for the quantitative determination of Fe(III). In Volhard’s method, it is used to determine the concentration of chloride ion by titration. It is also used as an analyzer for direct and continuous measurement of the concentration of soluble copper in an aqueous solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Odor Odorless
Appearance Colorless liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Information Miscible with water,acetone and alcohol.
Formula Weight 97.18
Chemical Name or Material Potassium thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.