missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 98%

Product Code. 11404754
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404754 500 g 500g
11414754 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404754 Supplier Thermo Scientific Alfa Aesar Supplier No. A13731.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 333-20-0 | CKNS | 97.176 g/mol

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

Solubility
Soluble in water, acetone and alcohol.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with nitric acids, active halogen compounds and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 172°C to 175°C
Density 1.89
pH 5.3 to 8.5
Boiling Point 500°C (decomposition)
Odor Odorless
Quantity 500 g
Beilstein 3594799
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,7691
Solubility Information Soluble in water,acetone and alcohol.
Formula Weight 97.18
Percent Purity 98%
Chemical Name or Material Potassium thiocyanate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Potassium thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.