missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 99+%, for analysis

Product Code. 10618313 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
2.5kg
250g
500g
This item is not returnable. View return policy

Product Code. 10618313

Brand: Thermo Scientific Acros 196580100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333-20-0 | CKNS | 97.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.18
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 170.0°C to 179.0°C
Color White
pH 5.3 to 8.7 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 10 kg
Assay Percent Range 99% min. (Argentometry)
Linear Formula KSCN
Fieser 01,954
Merck Index 15, 7807
Solubility Information Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 97.18
Percent Purity ≥99%
Grade Analytical
Packaging Plastic drum
Specific Gravity 1.886
Density 1.8860g/mL
Chemical Name or Material Potassium thiocyanate, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Potassium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.