missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Color | Colorless or White |
| Quantity | 500 g |
| Linear Formula | KSCN |
| UN Number | NONH for all modes of transport |
| Beilstein | 3594799 |
| Formula Weight | 97.18g/mol |
| Percent Purity | ≥99% |
| Grade | ACS Reagent |
| Chemical Name or Material | Potassium Thiocyanate |
Safety and Handling
Product Identifier
- Potassium thiocyanate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Potassium thiocyanate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction