missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10090310 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10090310 250 g 250g
10697222 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10090310 Supplier Fisher Chemical Supplier No. P/7280/50
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number 11413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 170°C
Color Colorless to White
pH 5.3 to 8.7
Physical Form Crystalline Solid
Quantity 250 g
Formula Weight 97.18g/mol
Packaging Plastic powder jar
Vapor Pressure <1 hPa at 20°C
compliance-icons
Product Identifier
  • Potassium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P233-Keep container tightly closed.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.