missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, TRC

Product Code. 30011393
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011393 500 g 500g
17974885 100 g 100g
17954635 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011393 Supplier Toronto Research Chemicals Supplier No. TRCP698520500G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.18
Synonym Thiocyanic Acid Potassium Salt, Arterocyn, Aterocyn, Kyonate, Potassium Isothiocyanate, Potassium Rhodanate, Potassium Thiocyanide, Rhocya, Rodanca, Thio-Cara
IUPAC Name potassium;thiocyanate
SMILES [K+].[S-]C#N

Specifications

Quantity 500 g
InChI Formula InChI=1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1
Formula Weight 96.9389
Chemical Name or Material Potassium Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.