missing translation for 'onlineSavingsMsg'
Learn More

Potassium trifluoroacetate, 98%

Product Code. 11443647
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11443647

Brand: Thermo Scientific Alfa Aesar A17398.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2923-16-2 | C2F3KO2 | 152.11 g/mol

It is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2923-16-2
Molecular Formula C2F3KO2
Molecular Weight (g/mol) 152.11
MDL Number MFCD00013215
InChI Key CUNPJFGIODEJLQ-UHFFFAOYSA-M
Synonym potassium trifluoroacetate, potassium 2,2,2-trifluoroacetate, trifluoroacetic acid potassium salt, acetic acid, trifluoro-, potassium salt, potassiumtrifluoroacetate, pubchem12605, potassium ion trifluoroacetate, ksc492g5t, trifluoroacetic acid, potassium salt, potassium 2,2,2-tris fluoranyl ethanoate
PubChem CID 23662811
SMILES [K+].[O-]C(=O)C(F)(F)F

Specifications

Melting Point 142°C to 145°C
pH 7
Quantity 100 g
UN Number UN2811
Beilstein 3717603
Sensitivity Hygroscopic
Solubility Information Soluble in water.
IUPAC Name potassium trifluoroacetate
Formula Weight 152.12
Percent Purity 98%
Chemical Name or Material Potassium trifluoroacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.