missing translation for 'onlineSavingsMsg'
Learn More

Potassium trifluoromethanesulfonate, 98%

Product Code. 11367947
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11367947

Brand: Thermo Scientific Alfa Aesar 020361.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2926-27-4 | CF3KO3S | 188.162 g/mol

Potassium trifluoromethanesulfonate is used as a reagent in the synthesis of guanine-quadruplex hybrid materials. It acts as a supporting electrolyte in the electrochemical study of evidence for gold anion in ethylenediamine. It is involved in the preparation of N-fluoro-2,4,6-trimethylpyridinium triflate by reacting with 2,4,6-trimethyl-pyridine. Further, it is used in the preparation of imidazolium salt, 3-methyl-1-(3R,3aR,6S,6aR)-[6-(benzyloxy)-hexahydrofuro[3,2-b]furan-3-yl]imidazolium trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2926-27-4
Molecular Formula CF3KO3S
Molecular Weight (g/mol) 188.162
MDL Number MFCD00042370
InChI Key GLGXXYFYZWQGEL-UHFFFAOYSA-M
Synonym potassium trifluoromethanesulfonate, potassium triflate, trifluoromethanesulfonic acid potassium salt, potassium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, potassium salt, potassium ion triflate, cf3so3k, acmc-209h7u, potassium trifluoromethanesulfonate salt, potassium tris fluoranyl methanesulfonate
PubChem CID 23664779
IUPAC Name potassium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[K+]

Specifications

Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 188.17
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Potassium trifluoromethanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.