missing translation for 'onlineSavingsMsg'
Learn More

Potassium trihydrogen dioxalate dihydrate, 98+%

Product Code. 11462757
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11462757

Brand: Thermo Scientific Alfa Aesar A14933.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol

Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-20-5
Molecular Formula C4H3KO8
Molecular Weight (g/mol) 218.16
MDL Number MFCD00150443,MFCD00150443
InChI Key GANDVAJEIJXBQJ-UHFFFAOYSA-M
Synonym potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
PubChem CID 131698588
SMILES [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O

Specifications

Quantity 1000 g
UN Number UN3288
Merck Index 14,7689
Solubility Information Soluble in water
IUPAC Name potassium;hydron;oxalate;dihydrate
Formula Weight 254.19 (218.16 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Potassium trihydrogen dioxalate dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.