missing translation for 'onlineSavingsMsg'
Learn More

Potassium trihydrogen dioxalate dihydrate, 98+%

Product Code. 11472757
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472757 250 g 250g
11462757 1000 g 1000g
11482757 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472757 Supplier Thermo Scientific Alfa Aesar Supplier No. A14933.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol

Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-20-5
Molecular Formula C4H3KO8
Molecular Weight (g/mol) 218.16
MDL Number MFCD00150443,MFCD00150443
InChI Key GANDVAJEIJXBQJ-UHFFFAOYSA-M
Synonym potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
PubChem CID 131698588
SMILES [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O

Specifications

Quantity 250 g
UN Number UN3288
Merck Index 14,7689
Solubility Information Soluble in water
IUPAC Name potassium oxalic acid hydrogen oxalate
Formula Weight 254.19 (218.16 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Potassium trihydrogen dioxalate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.