missing translation for 'onlineSavingsMsg'
Learn More

Potassium tris(1-pyrazolyl)borohydride, 93%

Product Code. 10572651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10572651 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10572651 Supplier Thermo Scientific Acros Supplier No. 296010050
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18583-60-3 | C9H10BKN6 | 252.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 18583-60-3
Melting Point 182°C to 188°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 93%
Packaging Glass bottle
Molecular Formula C9H10BKN6
MDL Number MFCD00040346
Quantity 5 g
Synonym potassium hydrotris 1-pyrazolyl borate, potassium hydridotri 1h-pyrazol-1-yl borate 1-, potassium hydro-tris 1-pyrazolyl borate, potassium tri 1h-pyrazol-1-yl hydroborate, potassium tri 1-pyrazolyl borohydride, potassium hydrotris 1-pyrazolato borate, hydrate
Solubility Information Solubility in water: reacts. Other solubilities: soluble in alcohols,thf,dmf
InChI Key IBNGZWIUPDMZMG-UHFFFAOYSA-N
SMILES [K+].C1=CN(N=C1)[BH-](N1C=CC=N1)N1C=CC=N1
IUPAC Name potassium tris(1H-pyrazol-1-yl)boranuide
Molecular Weight (g/mol) 252.13
Formula Weight 252.13
Percent Purity 93%
Physical Form Crystalline Powder
Chemical Name or Material Potassium tris(1-pyrazolyl)borohydride, 93%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.