missing translation for 'onlineSavingsMsg'
Learn More

Potassium vinyltrifluoroborate, 97%

Product Code. 11364855
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364855 1 g 1g
11374855 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364855 Supplier Thermo Scientific Alfa Aesar Supplier No. L17970.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13682-77-4 | C2H3BF3K | 133.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13682-77-4
Molecular Formula C2H3BF3K
Molecular Weight (g/mol) 133.95
MDL Number MFCD02093335
InChI Key ZCUMGICZWDOJEM-UHFFFAOYSA-N
Synonym potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
PubChem CID 23679353
IUPAC Name potassium;ethenyl(trifluoro)boranuide
SMILES [B-](C=C)(F)(F)F.[K+]

Specifications

Melting Point 230°C (decomposition)
Quantity 1 g
Beilstein 7777570
Sensitivity Hygroscopic
Formula Weight 133.95
Percent Purity 97%
Chemical Name or Material Potassium vinyltrifluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.