missing translation for 'onlineSavingsMsg'
Learn More

Potassium Vinyltrifluoroborate, TRC

Product Code. 30412020
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30412020 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30412020

Brand: Toronto Research Chemicals TRCP69870025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Vinyltrifluoroborate
CAS 13682-77-4
Molecular Formula C2H3BF3K
Quantity 25 g
Synonym Potassium Trifluorovinylborate, Potassium (Ethenyl)trifluoroborate, Potassium Vinyltrifluoroborate
InChI Formula InChI=1S/C2H3BF3.K/c1-2-3(4,5)6;/h2H,1H2;/q-1;+1
SMILES [K+].F[B-](F)(F)C=C
IUPAC Name potassium;ethenyl(trifluoro)boranuide
Molecular Weight (g/mol) 133.95
Formula Weight 133.9917
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.