missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Potato Dextrose Agar

Product Code. 11470225 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470225 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470225 Supplier MP Biomedicals™ Supplier No. 091008517
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the cultivation of yeasts and molds from dairy products and other foods.

Potato Dextrose Agar is used for the isolation, enumeration and identification of yeast and molds. It may also be used to induce sporulation in many fungi.

Specifications

Content And Storage Room Temperature
Description Potato Dextrose Agar
Media Type Yeast Media
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.