missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU5F1P1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | POU5F1P1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
POU5F1P1 Polyclonal antibody specifically detects POU5F1P1 in Human samples. It is validated for Western BlotSpecifications
| POU5F1P1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| OCT4-PG1, Octamer-binding protein 3-like, Octamer-binding transcription factor 3-like, OTF3Coctamer binding protein 3_like sequence, OTF3P1, POU 5 domain protein, POU class 5 homeobox 1 pseudogene 1, POU class 5 homeobox 1B, POU domain class 5, transcription factor 1 pseudogene 1, POU domain transcription factor Oct-4, POU domain transcription factor OCT4-pg1, POU domain, class 5, transcription factor 1 pseudogene 1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8, putative POU domain, class 5, transcription factor 1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human POU5F1B (NP_001153014.1). MAGHLASDFAFSPPPGGGGDGPWGAEPGWVDPLTWLSFQGPPGGPGIGPGVGPGSEVWGIPPCPPPYELCGGMAYCGPQVGVGLVPQGGLETSQPESEAGVGVESNSNGASPEPCTVPPGAVKLEKEKLEQNPEKSQDIK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 5462 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title