missing translation for 'onlineSavingsMsg'
Learn More

Povidone-iodine, TRC

Product Code. 17994385
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994385 250 g 250g
17902205 100 g 100g
17964805 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994385 Supplier Toronto Research Chemicals Supplier No. TRCP698900250G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25655-41-8
Molecular Formula (C6H9NO)n • [I2]x
Molecular Weight (g/mol) 366.97
Synonym 1-Ethenyl-2-pyrrolidinone Homopolymer compd. with Iodine, Iodine-poly(vinylpyrrolidone) complex, Betadine, Betaisodona, Betaseptic, Braunol, Braunosan H Disadine DP;
SMILES C=CN1CCCC1=O.II

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H9NO.I2/c1-2-7-5-3-4-6(7)8;1-2/h2H,1,3-5H2;
Formula Weight 366.893
Chemical Name or Material Povidone (iodinated)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.