missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Powder Flask

Product Code. 10045801 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
2000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10045801 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10045801 Supplier Heidolph™ Supplier No. 5147530000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with rotary evaporators

Powder flasks come with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Glass
Type Powder Flask
For Use With (Equipment) Rotary evaporators
Standard Taper Joint Number Center 29/32
Volume (Metric) 2000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.