missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marigold™ Powder-free Blue Nitrile Gloves
Click to view available options
:
Color: blue; Size: L
Specifications
Specifications
| Certifications/Compliance | CE 0086, 21 CFR (USA) and European Directives compliant for food applications, standards EN420 (General requirements), EN388 (Mechanical Hazards), EN374 (Chemical and Micro-organisms Hazards), EN421 (radioactive contamination only) by the notified laboratory BSI, Manufactured under EC quality assurance, system carried out by the BSI body |
| Classification | PPE Category III |
| Coating Material | Polymer |
| Color | Blue |
| Disposable | Single-use |
| Exterior Finish | Partially Textured |
| Hand | Ambidextrous |
| Material | Nitrile |
| Powdered | Powder Free |
| Product Type | Nitrile Examination Glove |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction