Learn More
Thermo Scientific™ Power Adapter for Eutech™ Meters
Click to view available options
For Use With (Equipment):
Cyberscan 11, 110, Eutech 700
Eutech 150, 450, 2700
Unit Size:
Each
Description
Replacement and optional adapters provide you with the power you need for your Eutech™ meters.
Features:
- Replacement and accessories for Eutech™ meters
Specifications
Specifications
| Description | 100 / 240 VAC SMPS power adapter, 9 V, 6 W |
| Type | Power Adapter |
| For Use With (Equipment) | Eutech 150, 450, 2700 |