Learn More
Thermo Scientific™ Power Adapter for Multiskan™ Ease
Product Code. 31921259 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
31921259 1 Ea. Each
1 options
Product Code. 31921259 Supplier Thermo Scientific™ Supplier No. MEAS0300
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Power Adapter for Multiskan Ease is the power supply for the Multiskan™ Ease Absorbance Microplate Reader, a modern easy-to-use absorbance reader designed to simplify both routine and advanced microplate-based assays.
The Power Adapter for Multiskan Ease is the power supply for the Multiskan™ Ease Absorbance Microplate Reader, a modern easy-to-use absorbance reader designed to simplify both routine and advanced microplate-based assays. It enables consistent performance and safe operation in laboratory environments. Designed for global use, it features a universal input range of 100–240 VAC (50/60 Hz, 1.4 A) and enables a stable 24 V, 5 A output with a maximum power of 120 W.
Key Features
  • Built-in protection against short circuit, overload, over-voltage, and over-temperature
  • Fully enclosed, durable housing suitable for lab use
  • Certified to IEC / EN / UL 62368-1 standards
  • CE and UKCA marked
  • FCC, ICES, and RoHS compliant
This power adapter is specifically compatible with the Thermo Scientific™ Multiskan™ Ease Absorbance Microplate Reader. Its robust construction supports long-term use in demanding laboratory settings, while comprehensive international safety and regulatory certifications help compliance and operational confidence across global markets.
TRUSTED_SUSTAINABILITY
Specifications
Product Type External Power
For Use With (Equipment) Multiskan Ease
Quantity 1 Ea.