missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Power Cord

Product Code. 31906277 Shop All Thermo Scientific Products
Change view
Click to view available options
Connection Type:
EU
Length (Metric):
1.8 m
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Length (Metric) Unit Size
31906277 EU 1.8 m Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31906277 Supplier Thermo Scientific Supplier No. 6000.1000
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Power cord accessory for compatible regional power connections.

  • Accessory available in EU, UK, Swiss, and Denmark versions to support regional power connection requirements.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Gray
Connection Type EU
For Use With (Equipment) All HPLC and UHPLC Systems
Type Cable Assembly
Disposable Reusable
Length (Metric) 1.8 m
Voltage 230 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.